Liquiritin

Lieferant: Thermo Fisher Scientific

465482500
ACRO465482500EA 372 EUR
ACRO465482500
Liquiritin
Liquiritin

  • Bioactive

Formel: C₂₁H₂₂O₉
Molecular Weight: 418.4 g/mol
Schmelzpunkt: >207 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00210526
CAS-Nummer: 551-15-5

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤4 %
Solubility (1 mg/ml in EtOH) Clear colorless
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR