5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5,5'-Diallylbiphenyl-2,2'-diol, 5,5'-Diallyl-[1,1'-biphenyl]-2,2'-diol, Magnolol

465512500
ACRO465512500EA 80.3 EUR
ACRO465512500
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)
Formel: C₁₈H₁₈O₂
Molecular Weight: 266,34 g/mol
Schmelzpunkt: 101°C…102°C
Lagertemperatur: Kühlschrank
MDL: MFCD00016658
CAS-Nummer: 528-43-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige powder
Infrared spectrum Conforms
Melting point 99°C to 103°C
Water ≤ 1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR