Pixantrone maleate

Lieferant: Thermo Scientific
Achtung

Synonyme: Pixantrone, 6,9-Bis((2-aminoethyl)amino)benzo[g]isoquinoline-5,10-dione dimaleate

465522500
ACRO465522500EA 232 EUR
ACRO465522500
Pixantrone maleate
Pixantrone maleate
Formel: C₁₇H₁₉N₅O₂·C₈H₈O₈
Molecular Weight: 557,52 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00920108
CAS-Nummer: 144675-97-8

Bestellen

Spezifikation Testergebnisse

Appearance Purple to brown to black powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Solubility (2 mg/ml in water) Clear solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR