Valsartan
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid
465560010
465560050
ACRO465560010EA
32.4
EUR
ACRO465560010
ACRO465560050
Valsartan
Valsartan
Formel:
C₂₄H₂₉N₅O₃ Molecular Weight: 435.52 g/mol Schmelzpunkt: 117°C Lagertemperatur: Kühlschrank |
MDL:
MFCD00865840 CAS-Nummer: 137862-53-4 Merck Index: 13.9982 |
Spezifikation Testergebnisse
Appearance | White to almost white to light beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 97.5% |
Specific optical rotation | -55° to -70° (20°C, 589nm) (1% in CH3OH) |
Water | ≤ 2% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...