Sulphamethazine sodium salt

Lieferant: Thermo Fisher Scientific
Achtung

465600050
ACRO465600050EA 17.8 EUR
ACRO465600050
Sulphamethazine sodium salt
Sulphamethazine sodium salt
Formel: C₁₂H₁₃N₄NaO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD00068333
CAS-Nummer: 1981-58-4
EINECS: 217-840-0

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
Loss on drying ≤ 2%
HPLC ≥ 98.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR