Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid)

Lieferant: Thermo Scientific
Achtung

Synonyme: Acridine-3,6-diamine hydrochloride

465860050
ACRO465860050EA 39.5 EUR
ACRO465860050
Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid)
Proflavin Hydrochlorid (3,6-Diaminoacridin Hydrochlorid)
Formel: C₁₃H₁₁N₃·HCl
Molecular Weight: 245,71 g/mol
Schmelzpunkt: 270°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066867
CAS-Nummer: 952-23-8
EINECS: 213-469-9

Bestellen

Spezifikation Testergebnisse

Appearance Dark orange to red-brown powder
Infrared spectrum Conforms
Titration Argentometry 13.6 to 15.0% (Cl)
HPLC ≥ 94%
Loss on drying ≤ 4% (105°C, 1hr)
Solubility (1mg/ml in water) Clear solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR