Gemifloxacin mesylate

Lieferant: Thermo Fisher Scientific

Synonyme: (Z)-7-(3-(Aminomethyl)-4-(methoxyimino)pyrrolidin-1-yl)-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid methanesulphonic acid salt

465972500
ACRO465972500EA 253 EUR
ACRO465972500
Gemifloxacin mesylate
Gemifloxacin mesylate
Formel: C₁₈H₂₀FN₅O₄·CH₄O₃S
Lagertemperatur: Kühlschrank
MDL: MFCD08277637
CAS-Nummer: 210353-53-0

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Water ≤ 10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR