Cinacalcet hydrochloride
Lieferant: Thermo Scientific
Synonyme:
(R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propan-1-amine hydrochloride
466010010
ACRO466010010EA
155
EUR
ACRO466010010
Cinacalcet hydrochloride
Cinacalcet hydrochloride
Spezifikation Testergebnisse
Appearance | White to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥ 98.0% |
Water | ≤ 0.5% |
Specific optical rotation | -32.0° to -24.0° (20°C, 589nm) (c=0.5 in methanol) |
Solubility | (2mg/ml in DMSO) Clear |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...