Nelfinavir mesylate
Lieferant: Thermo Scientific
Synonyme:
(3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide methanesulphonate, AG 1341
466200010
ACRO466200010EA
1758
EUR
ACRO466200010
Nelfinavir mesylate
Nelfinavir mesylate
Formel:
C₃₂H₄₅N₃O₄S·CH₄O₃S Molecular Weight: 663,9 g/mol Schmelzpunkt: 131°C…135°C Lagertemperatur: Kühlschrank |
MDL:
MFCD00931436 CAS-Nummer: 159989-65-8 Merck Index: 13.6471 |
Spezifikation Testergebnisse
Appearance | White to light brown powder |
Infrared spectrum | Conforms |
HPLC | ≥ 96.0% |
Specific optical rotation | -109° to -115° (20°C, 589nm) (1% in Methanol) (on dry substance) |
Water | 2 to 8% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...