Mildronate dihydrate

Lieferant: Thermo Scientific
Achtung

466260010
ACRO466260010EA 158 EUR
ACRO466260010
Mildronate dihydrate
Mildronate dihydrate
Formel: C₆H₁₄N₂O₂·₂H₂O
Molecular Weight: 182,22 g/mol
Schmelzpunkt: 85°C…90°C
Lagertemperatur: Kühlschrank
MDL: MFCD00876389
CAS-Nummer: 86426-17-7

Bestellen

Spezifikation Testergebnisse

Appearance White to beige crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5%
Water 18 to 22%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR