1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%

Lieferant: Thermo Fisher Scientific
Achtung

466330050 466330250
ACRO466330050EA 50.2 EUR
ACRO466330050 ACRO466330250
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%
1-(tert-Butoxycarbonyl)-2-pyrrolidinone
Formel: C₉H₁₅NO₃
Dichte: 1,086 g/mL (25 °C) (Lit.)
MDL: MFCD00674099
CAS-Nummer: 85909-08-6

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Clear colorless to yellow to brown
Appearance (Form) Liquid
Infrared spectrum Conforms
Refractive index 1.4645 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR