Flutamid

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(4-Nitro-3-(trifluoromethyl)phenyl)isobutyramide, 4'-Nitro-3'-(trifluoromethyl)isobutyranilide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide

466450050
ACRO466450050EA 79.4 EUR
ACRO466450050
Flutamid
Flutamide
Formel: C₁₁H₁₁F₃N₂O₃
Molecular Weight: 276,22 g/mol
Schmelzpunkt: 112°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00072009
CAS-Nummer: 13311-84-7
EINECS: 236-341-9
Merck Index: 13.4234

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow powder
Infrared spectrum Conforms
Melting point 110.0°C to 114.0°C
HPLC ≥ 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR