Matrin

Lieferant: Thermo Scientific
Achtung

Synonyme: (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, Matridin-15-one

466592500
ACRO466592500EA 94 EUR
ACRO466592500
Matrin
Matrin
Formel: C₁₅H₂₄N₂O
Molecular Weight: 248,37 g/mol
Schmelzpunkt: 76°C
Lagertemperatur: Kühlschrank
MDL: MFCD00210527
CAS-Nummer: 519-02-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow powder
Infrared spectrum Conforms
Water ≤ 5%
GC ≥ 97.0%
Specific optical rotation +31.0° to +36.0° (20°C, 589nm) (c=1, EtOH, dry substance)
Solubility (5% water) Clear colourless to pale yellow solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR