Mephenytoin
Lieferant: Thermo Fisher Scientific
|
Achtung
|
466690250
ACRO466690250EA
127
EUR
ACRO466690250
Mephenytoin
Mephenytoin
Formel:
C₁₂H₁₄N₂O₂ Schmelzpunkt: 135°C…137°C Lagertemperatur: Kühlschrank |
MDL:
MFCD00022407 CAS-Nummer: 50-12-4 EINECS: 200-012-8 Merck Index: 13.5878 |
Spezifikation Testergebnisse
Appearance | White to almost white crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥ 98.0% (sum of isomers) |
Water | ≤ 2.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...