Abiraterone Acetate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate
466730010
ACRO466730010EA
251
EUR
ACRO466730010
Abiraterone Acetate
Abiraterone Acetate
Formel:
C₂₆H₃₃NO₂ Molecular Weight: 391,56 g/mol Schmelzpunkt: 147°C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00934213 CAS-Nummer: 154229-18-2 |
Spezifikation Testergebnisse
Appearance | White to almost white crystalline powder |
Infrared spectrum | Conforms |
Melting point | 143.0°C to 147.0°C |
HPLC | ≥ 98.0% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...