Temozolomide

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide

466760010
ACRO466760010EA 97.8 EUR
ACRO466760010
Temozolomide
Temozolomide
Formel: C₆H₆N₆O₂
Molecular Weight: 194.15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00866492
CAS-Nummer: 85622-93-1
Merck Index: 13.9218

Bestellen

Spezifikation Testergebnisse

Appearance White to light pink to brown powder
Infrared spectrum Conforms
Loss on drying ≤ 1%
HPLC ≥ 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR