Idebenon

Lieferant: Thermo Scientific

Synonyme: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione

466770010
ACRO466770010EA 67.4 EUR
ACRO466770010
Idebenon
Idebenone
Formel: C₁₉H₃₀O₅
Molecular Weight: 338,44 g/mol
Schmelzpunkt: 52°C…55°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00274552
CAS-Nummer: 58186-27-9
Merck Index: 13.4913

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange powder
Infrared spectrum Conforms
Melting point 51.0°C to 55.0°C
HPLC ≥ 98.0%
Loss on drying ≤ 0.5%
Solubility (5% in methanol) Clear to slight yellow to orange solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR