Ibrutinib

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (R)-1-(3-(4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)prop-2-en-1-one, PCI-32765

466802500
ACRO466802500EA 56.1 EUR
ACRO466802500
Ibrutinib
Ibrutinib
Formel: C₂₅H₂₄N₆O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD20261150
CAS-Nummer: 936563-96-1

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 98.0%
Water ≤ 1.0%
Solubility (0.5% in DMSO) Clear colorless solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR