Gentiopicroside
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin
466832500
ACRO466832500EA
185
EUR
ACRO466832500
Gentiopicroside
Gentiopicroside
Formel:
C₁₆H₂₀O₉ Molecular Weight: 356,33 g/mol Schmelzpunkt: 191°C Lagertemperatur: Kühlschrank |
MDL:
MFCD00075700 CAS-Nummer: 20831-76-9 EINECS: 244-070-2 Merck Index: 13.4408 |
Spezifikation Testergebnisse
Appearance | White to light yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥ 98.0% |
Water | ≤ 2.0% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...