Teniposide

Lieferant: Thermo Scientific
Gefahr

Synonyme: (5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one

466852500
ACRO466852500EA 155 EUR
ACRO466852500
Teniposide
Teniposide
Formel: C₃₂H₃₂O₁₃S
Molecular Weight: 656,66 g/mol
Schmelzpunkt: 242°C…246°C
Lagertemperatur: Gefrierschrank
MDL: MFCD00866516
CAS-Nummer: 29767-20-2
EINECS: 249-831-2
Merck Index: 13.9222

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 96.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR