Azetidine-2-carboxylic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

466900010 466900050
ACRO466900010EA 89.4 EUR
ACRO466900010 ACRO466900050
Azetidine-2-carboxylic acid 97%
Azetidine-2-carboxylic acid
Formel: C₄H₇NO₂
MDL: MFCD00066660
CAS-Nummer: 2517-04-6

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to almost white
Appearance (Form) Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR