Azetidine-2-carboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
466900010
466900050
ACRO466900010EA
89.4
EUR
ACRO466900010
ACRO466900050
Azetidine-2-carboxylic acid 97%
Azetidine-2-carboxylic acid
Formel:
C₄H₇NO₂ |
MDL:
MFCD00066660 CAS-Nummer: 2517-04-6 |
Spezifikation Testergebnisse
Appearance (Color) | White to almost white |
Appearance (Form) | Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...