Desonide

Lieferant: Thermo Scientific

Synonyme: (11beta,16alpha)-11,21-Dihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Budesonide Acetonide

467450010
ACRO467450010EA 53.6 EUR
ACRO467450010
Desonide
Desonide
Formel: C₂₄H₃₂O₆
Molecular Weight: 416,51 g/mol
Schmelzpunkt: 275°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00866151
CAS-Nummer: 638-94-8
EINECS: 211-351-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥ 97.5%
Loss on drying ≤ 0.5% (1g, 105°C, 3hr)
Specific optical rotation +104° to +110° (20°C, 589nm) (10 mg/ml in dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR