Metamizole sodium monohydrate

Lieferant: Thermo Scientific
Achtung

467482500
ACRO467482500EA 93.9 EUR
ACRO467482500
Metamizole sodium monohydrate
Metamizole sodium monohydrate
Formel: C₁₃H₁₆N₃NaO₄S·H₂O
Molecular Weight: 351,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150293
CAS-Nummer: 5907-38-0
Merck Index: 13.3385

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light beige powder or crystals
Infrared spectrum Conforms
Melting point 211°C to 216°C
HPLC ≥ 97.5%
Water 4.5 to 5.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR