Cimetidin
Lieferant: Thermo Scientific
Synonyme:
2-Cyano-1-methyl-3-[2-(5-methyl-1H-imidazol-4-yl-methylthio)ethyl]guanidine, N-Cyano-N'-methyl-N''-[2-[[(4-methyl-1H-imidazol-5-yl)methyl]thio]ethyl]guanidine
467520050
ACRO467520050EA
46.7
EUR
ACRO467520050
Cimetidin
Cimetidine
Formel:
C₁₀H₁₆N₆S Molecular Weight: 252,34 g/mol Schmelzpunkt: 141°C…143°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01724314 CAS-Nummer: 51481-61-9 EINECS: 257-232-2 Merck Index: 13.23 |
Spezifikation Testergebnisse
Appearance | White to almost white crystalline powder |
Infrared spectrum | Conforms |
Melting point | 141°C to 143°C |
HPLC | ≥ 97.5% |
Loss on drying | ≤ 0.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...