Posaconazole
Lieferant: Thermo Scientific
|
Achtung
|
467660010
ACRO467660010EA
112
EUR
ACRO467660010
Posaconazole
Posaconazole
Formel:
C₃₇H₄₂F₂N₈O₄ Molecular Weight: 700,79 g/mol Schmelzpunkt: 170°C…179°C Lagertemperatur: Kühlschrank |
MDL:
MFCD00941162 CAS-Nummer: 171228-49-2 |
Spezifikation Testergebnisse
Appearance | White to light yellow to beige powder or crystals |
Infrared spectrum | Conforms |
Melting point | 165°C to 170°C |
Water | ≤ 1% |
Specific optical rotation | -24° to -30° (20°C, 589nm) (1% in CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...