AGI-6780 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide

467821000
ACRO467821000EA 404 EUR
ACRO467821000
AGI-6780 95%
AGI-6780

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₁H₁₈F₃N₃O₃S₂
Molecular Weight: 481.51 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26097285
CAS-Nummer: 1432660-47-3

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR