I-BET151 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 7-(3,5-Dimethylisoxazol-4-yl)-8-methoxy-1-((R)-1-(pyridin-2-yl)ethyl)-1H-imidazo[4,5-c]quinolin-2(3H)-one

467841000
ACRO467841000EA 564 EUR
ACRO467841000
I-BET151 95%
I-BET151

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₃H₂₁N₅O₃
Molecular Weight: 415,4445 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD22124472
CAS-Nummer: 1300031-49-5

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to light brown Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR