JW55 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(4-(((4-(4-Methoxyphenyl)tetrahydro-2H-pyran-4-yl)methyl)carbamoyl)phenyl)furan-2-carboxamide, N-[4-[[[[Tetrahydro-4-(4-methoxyphenyl)-2H-pyran-4-yl]methyl]amino]carbonyl]phenyl]-2-furancarboxamide

467851000
ACRO467851000EA 440 EUR
ACRO467851000
JW55 95%
JW55

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₅H₂₆N₂O₅
Molecular Weight: 434,48 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03863265
CAS-Nummer: 664993-53-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR