GDC-0941 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(2-(1H-Indazol-4-yl)-6-((4-(methylsulphonyl)piperazin-1-yl)methyl)thieno[3,2-d]pyrimidin-4-yl)morpholine, 4-[2-(1H-Indazol-4-yl)-6-[(4-methylsulphonylpiperazin-1-yl)methyl]thieno[3,2-d]pyrimidin-4-yl]morpholine, Pictilisib
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.
Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies
Formel:
C₂₃H₂₇N₇O₃S₂ Molecular Weight: 513,6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11616196 CAS-Nummer: 957054-30-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or solid |
HPLC | ≥94% |
MS | complies |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...