UNC-2025 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: MRX-6313, (1R,4R)-4-(2-(butylamino)-5-(4-((4-methylpiperazin-1-yl)methyl)phenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol, UNC2025, trans-4-(2-(Butylamino)-5-(4-((4-methylpiperazin-1-yl)methyl)phenyl)-7H-pyrrolo[2,3-d]pyrimidin-7-yl)cyclohexanol

467881000
ACRO467881000EA 448 EUR
ACRO467881000
UNC-2025 95%
UNC-2025

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₈H₄₀N₆O
Molecular Weight: 476,66 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28142874
CAS-Nummer: 1429881-91-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR