VE-821 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-6-(4-(methylsulphonyl)phenyl)-N-phenylpyrazine-2-carboxamide, VE 821, VE-821, VE821, 3-amino-6-(4-methylsulphonylphenyl)-N-phenylpyrazine-2-carboxamide

467891000
ACRO467891000EA 219 EUR
ACRO467891000
VE-821 95%
VE-821

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₁₈H₁₆N₄O₃S
Molecular Weight: 368,41 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD19443686
CAS-Nummer: 1232410-49-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR