A-1210477 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 7-(5-((4-(4-(N,N-dimethylsulphamoyl)piperazin-1-yl)phenoxy)methyl)-1,3-dimethyl-1H-pyrazol-4-yl)-1-(2-morpholinoethyl)-3-(3-(naphthalen-1-yloxy)propyl)-1H-indole-2-carboxylic acid

467901000
ACRO467901000EA 325 EUR
ACRO467901000
A-1210477 95%
A-1210477

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₄₆H₅₅N₇O₇S
Molecular Weight: 850,04 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28399118
CAS-Nummer: 1668553-26-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR