ACY-738 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Hydroxy-2-((1-phenylcyclopropyl)amino)pyrimidine-5-carboxamide

467911000
ACRO467911000EA 345 EUR
ACRO467911000
ACY-738 95%
ACY-738

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₁₄H₁₄N₄O₂
Molecular Weight: 270,29 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28347706
CAS-Nummer: 1375465-91-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR