Bafetinib 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-N-(3-([4,5'-Bipyrimidin]-2-ylamino)-4-methylphenyl)-4-((3-(dimethylamino)pyrrolidin-1-yl)methyl)-3-(trifluoromethyl)benzamide, NS-187

467941000
ACRO467941000EA 640 EUR
ACRO467941000
Bafetinib 95%
Bafetinib

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₀H₃₁F₃N₈O
Molecular Weight: 576,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD18633200
CAS-Nummer: 859212-16-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light brown Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR