AZ191 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-[2-Methoxy-4-(4-methyl-1-piperazinyl)phenyl]-4-(1-methyl-1H-pyrrolo[2,3-c]pyridin-3-yl)-2-pyrimidinamine

467951000
ACRO467951000EA 640 EUR
ACRO467951000
AZ191 95%
AZ191

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₄H₂₇N₇O
Molecular Weight: 429.52 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28015098
CAS-Nummer: 1594092-37-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR