(R)-BI-2536 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
BI-2536, BI 2536, (R)-4-((8-Cyclopentyl-7-ethyl-5-methyl-6-oxo-5,6,7,8-tetrahydropteridin-2-yl)amino)-3-methoxy-N-(1-methylpiperidin-4-yl)benzamide
Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.
Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies
Formel:
C₂₈H₃₉N₇O₃ Molecular Weight: 521,7 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD10565924 CAS-Nummer: 755038-02-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder or solid |
HPLC | ≥94% |
MS | complies |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...