BIX2188 Z-Isomer 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
BIX-02188, BIX02188, (Z)-3-(((3-((dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide
467971000
ACRO467971000EA
440
EUR
ACRO467971000
BIX2188 Z-Isomer 95%
BIX2188 Z-Isomer
Chemical probes can be used to help establish the relationship between a molecular target and the broader biological consequences of modulating that target in cells or organisms.
Chemical probes are selective small-molecule modulators of a protein’s function allowing the user to ask mechanistic and phenotypic questions about their molecular target in cell-based or animal studies
Formel:
C₂₅H₂₄N₄O₂ Molecular Weight: 412,493 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD18157583 CAS-Nummer: 334949-59-6 |
Spezifikation Testergebnisse
Appearance | White to yellow Powder or solid |
HPLC | ≥94% |
MS | complies |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...