CFI-400945 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (1R,2S)-2-(3-((E)-4-(((2R,6S)-2,6-Dimethylmorpholino)methyl)styryl)-1H-indazol-6-yl)-5'-methoxyspiro[cyclopropane-1,3'-indolin]-2'-one, (1R,2S)-2-(3-((E)-4-(((2S,6R)-2,6-dimethylmorpholino)methyl)styryl)-1H-indazol-6-yl)-5'-methoxyspiro[cyclopropane-1,3'-indolin]-2'-one

467981000
ACRO467981000EA 648 EUR
ACRO467981000
CFI-400945 95%
CFI-400945

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₃H₃₄N₄O₃
Molecular Weight: 534,66 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28386170
CAS-Nummer: 1338806-73-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR