CP-673451 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(2-(5-(2-Methoxyethoxy)-1H-benzo[d]imidazol-1-yl)quinolin-8-yl)piperidin-4-amine, 1-[2-[5-(2-methoxyethoxy)benzimidazol-1-yl]quinolin-8-yl]piperidin-4-amine

467991000
ACRO467991000EA 539 EUR
ACRO467991000
CP-673451 95%
CP-673451

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₄H₂₇N₅O₂
Molecular Weight: 417,51 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11100329
CAS-Nummer: 343787-29-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR