GW-3965 hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: GW3965 hydrochloride, 2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)propoxy)phenyl)acetic acid hydrochloride, 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2,2-diphenylethyl)amino]propoxy]benzeneacetic acid hydrochloride

468011000
ACRO468011000EA 320 EUR
ACRO468011000
GW-3965 hydrochloride 95%
GW-3965 hydrochloride

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₃H₃₁ClF₃NO₃·HCl
Molecular Weight: 618,51 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08276920
CAS-Nummer: 405911-17-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR