GNE-7915 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-((4-(Ethylamino)-5-(trifluoromethyl)pyrimidin-2-yl)amino)-2-fluoro-5-methoxyphenyl)(morpholino)methanone, [4-[[4-(ethylamino)-5-(trifluoromethyl)pyrimidin-2-yl]amino]-2-fluoro-5-methoxyphenyl]-morpholin-4-ylmethanone

468021000
ACRO468021000EA 409 EUR
ACRO468021000
GNE-7915 95%
GNE-7915

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₁₉H₂₁F₄N₅O₃
Molecular Weight: 443.4 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD25977005
CAS-Nummer: 1351761-44-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR