GSK-2801 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: GSK2801, 1-(1-(2-(Methylsulphonyl)phenyl)-7-propoxyindolizin-3-yl)ethanone

468031000
ACRO468031000EA 555 EUR
ACRO468031000
GSK-2801 95%
GSK-2801

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₀H₂₁NO₄S
Molecular Weight: 371,54 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD26142953
CAS-Nummer: 1619994-68-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR