GSK-5959 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-piperidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide, GSK5959

468041000
ACRO468041000EA 406 EUR
ACRO468041000
GSK-5959 95%
GSK-5959

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₂H₂₆N₄O₃
Molecular Weight: 394,47 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 901245-65-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR