A-366 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5'-Methoxy-6'-[3-(1-pyrrolidinyl)propoxy]spiro[cyclobutane-1,3'-indol]-2'-amin, 5'-Methoxy-6'-[3-(1-pyrrolidinyl)propoxy]spiro[cyclobutane-1,3'-[3H]indol]-2'-amine, 5'-methoxy-6'-[3-(pyrrolidin-1-yl)propoxy]spiro[cyclobutane-1,3'-indole]-2'-amine, 5'-Methoxy-6'-(3-(pyrrolidin-1-yl)propoxy)spiro[cyclobutane-1,3'-indol]-2'-amine
Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.
Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies
Formel:
C₁₉H₂₇N₃O₂ Molecular Weight: 329,44 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD28133403 CAS-Nummer: 1527503-11-2 |
Spezifikation Testergebnisse
Appearance | Yellow to green to light brown Solid |
HPLC | ≥94% |
MS | complies |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...