CH5424802 (Alectinib) 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: Alectinib, 9-Ethyl-6,6-dimethyl-8-(4-morpholinopiperidin-1-yl)-11-oxo-6,11-dihydro-5H-benzo[b]carbazole-3-carbonitrile, 9-Ethyl-6,6-dimethyl-8-(4-morpholin-4-ylpiperidin-1-yl)-11-oxo-5H-benzo[b]carbazol-3-carbonitrile

468081000
ACRO468081000EA 502 EUR
ACRO468081000
CH5424802 (Alectinib) 95%
CH5424802 (Alectinib)

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₃₀H₃₄N₄O₂
Molecular Weight: 482,62 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD19440988
CAS-Nummer: 1256580-46-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR