AZD1152-HQPA 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(5-((7-(3-(Ethyl(2-hydroxyethyl)amino)propoxy)quinazolin-4-yl)amino)-1H-pyrazol-3-yl)-N-(3-fluorophenyl)acetamide, Barasertib-HQPA, Barasertib

468091000
ACRO468091000EA 282 EUR
ACRO468091000
AZD1152-HQPA 95%
AZD1152-HQPA

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₆H₃₀FN₇O₃
Molecular Weight: 507,56 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD17392583
CAS-Nummer: 722544-51-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR