BLU9931 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(2-((6-(2,6-Dichloro-3,5-dimethoxyphenyl)quinazolin-2-yl)amino)-3-methylphenyl)acrylamide

468111000
ACRO468111000EA 413 EUR
ACRO468111000
BLU9931 95%
BLU9931

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₆H₂₂Cl₂N₄O₃
Molecular Weight: 509.38 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28900729
CAS-Nummer: 1538604-68-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR