Alpelisib 95%

Lieferant: Thermo Fisher Scientific
Achtung

468121000
ACRO468121000EA 661 EUR
ACRO468121000
Alpelisib 95%
Alpelisib

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₁₉H₂₂F₃N₅O₂S
Lagertemperatur: Gefrierschrank
MDL: MFCD22417085
CAS-Nummer: 1217486-61-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder or crystalline powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR