EI1 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Cyano-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-1-(pentan-3-yl)-1H-indole-4-carboxamide, 6-cyano-N-[(1,2-dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-1-(1-ethylpropyl)-1H-indole-4-carboxamide

468151000
ACRO468151000EA 591 EUR
ACRO468151000
EI1 95%
EI1

Chemical probes can be used to help establish the
relationship between a molecular target and the broader
biological consequences of modulating that target in cells
or organisms.


Chemical probes are selective small-molecule
modulators of a protein’s function allowing the user to
ask mechanistic and phenotypic questions about their
molecular target in cell-based or animal studies

Formel: C₂₃H₂₆N₄O₂
Molecular Weight: 390,5 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD25976740
CAS-Nummer: 1418308-27-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or solid
HPLC ≥94%
MS complies
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR